Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-HLA-A Mouse Monoclonal Antibody (CF488A) [clone: 108-2C5]
Anti-HLA-A Mouse Monoclonal Antibody (CF488A) [clone: 108-2C5]
Supplier:  Biotium
Certificates
Anti-HLA-A Mouse Monoclonal Antibody (CF488A) [clone: 108-2C5]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75960-762
  • 75960-760
  • Antigen Symbol:
  • HLA-A
  • HLA-A
  • Antigen Name:
  • MHC I
  • MHC I
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • 108-2C5
  • 108-2C5
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 181244; 654404; 77961
  • Unigene ID: 181244; 654404; 77961
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together