Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-KRT Mouse Monoclonal Antibody (Biotin) [clone: KRT7/760 / KRT17/ 778 / KRT18/834 / KRTL/828 / KRTH/829 / KRT/457]
Anti-KRT Mouse Monoclonal Antibody (Biotin) [clone: KRT7/760 / KRT17/ 778 / KRT18/834 / KRTL/828 / KRTH/829 / KRT/457]
Supplier:  Biotium
Certificates
Anti-KRT Mouse Monoclonal Antibody (Biotin) [clone: KRT7/760 / KRT17/ 778 / KRT18/834 / KRTL/828 / KRTH/829 / KRT/457]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75968-018
  • 75968-016
  • Antigen Symbol:
  • KRT
  • KRT
  • Antigen Name:
  • Cytokeratin
  • Cytokeratin
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • KRT7/760 / KRT17/ 778 / KRT18/834 / KRTL/828 / KRTH/829 / KRT/457
  • KRT7/760 / KRT17/ 778 / KRT18/834 / KRTL/828 / KRTH/829 / KRT/457
  • Reactivity:
  • Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
  • Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 334989 (KRT77) , 654392 (KRT76)
  • Unigene ID: 334989 (KRT77) , 654392 (KRT76)
  • Isotype:
  • IgGs
  • IgGs
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications