Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CDC2 Mouse Monoclonal Antibody (CF640R) [clone: CDK1/873]
Anti-CDC2 Mouse Monoclonal Antibody (CF640R) [clone: CDK1/873]
Supplier:  Biotium
Certificates
Anti-CDC2 Mouse Monoclonal Antibody (CF640R) [clone: CDK1/873]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75967-440
  • 75969-034
  • Antigen Symbol:
  • CDC2
  • CDC2
  • Antigen Name:
  • p34 / cdk1
  • p34 / cdk1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • CDK1/873
  • CDK1/873
  • Reactivity:
  • Monkey, Human, Bovine, Mink
  • Monkey, Human, Bovine, Mink
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 334562
  • Unigene ID: 334562
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/ml
  • 0.1 mg/ml
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together