Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-RBP1 Mouse Monoclonal Antibody (CF488A) [clone: RBP/872]
 :  Biotium
 
Anti-RBP1 Mouse Monoclonal Antibody (CF488A) [clone: RBP/872]
 :  Biotium

 

  • Catalog No:
  • 75972-954
  • 75972-952
  • Antigen Symbol:
  • RBP1
  • RBP1
  • Antigen Name:
  • Retinol Binding Protein-1
  • Retinol Binding Protein-1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • RBP/872
  • RBP/872
  • Reactivity:
  • Rabbit, Monkey, Goat, Human, Rat, Mouse
  • Rabbit, Monkey, Goat, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 529571
  • Unigene ID: 529571
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide