Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-Golgi Complex Mouse Monoclonal Antibody (CF488A) [clone: AE-6]
 :  Biotium
 
Anti-Golgi Complex Mouse Monoclonal Antibody (CF488A) [clone: AE-6]
 :  Biotium

 

  • Catalog No:
  • 75949-042
  • 75949-044
  • Antigen Name:
  • Golgi Complex
  • Golgi Complex
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • AE-6
  • AE-6
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

 

Frequently Bought Together