About this item
This MAb reacts with a protein of 20-30 kDa, identified as PGP 9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UCHL-1). Initially, PGP 9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP 9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP 9.5 gene is believed to cause a form of Parkinson's disease.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.
Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: Clone UCHL1/841
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Specifications
- Catalog No:
- 75969-974
- 75969-972
- Antigen Symbol:
- UCHL1
- UCHL1
- Antigen Name:
- pgp9.5
- pgp9.5
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- UCHL1/841
- UCHL1/841
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 518731
- Unigene ID: 518731
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western Blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.1 mg/ml
- 0.1 mg/ml
- Storage Buffer:
- PBS/0.1% BSA/0.05% azide
- PBS/0.1% BSA/0.05% azide