Specifications
- Antibody Type:Primary
- Antigen Name:CD5
- Antigen Symbol:CD5
- Clonality:Monoclonal
- Clone:C5/473 CD5/54/F6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgGs
- Reactivity:Rabbit,Human,Bovine,Rat
- Size:50 μL
- Gene ID:Unigene ID: 58685
- Storage Buffer:PBS, no BSA, no azide
- Concentration:1 mg/mL
- Cat. No.:75911-892
- Supplier no.:BNUM0748-50
Specifications
About this item
This antibody recognizes a 67 kDa transmembrane protein, which is identified as CD5. The CD5 antigen is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. Anti-CD5 is a pan T-cell marker that also reacts with a range of neoplastic B-cells, e.g. chronic lymphocytic leukemia/small lymphocytic lymphoma (CLL/SLL), mantle cell lymphoma, and a subset (~10%) of diffuse large B-cell lymphoma. CD5 aberrant expression is useful in making a diagnosis of mature T-cell neoplasms. Anti-CD5 detection is diagnostic in CLL/SLL within a panel of other B-cell markers, especially one that includes anti-CD23. Anti-CD5 is also very useful in differentiating among mature small lymphoid cell malignancies. In addition, anti-CD5 can be used in distinguishing thymic carcinoma ( ) from thymoma (-). Anti-CD5 does not react with granulocytes or monocytes.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: Clone C5/473 CD5/54/F6
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Cow, Human, Rabbit, Rat