Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ERBB2 Mouse Monoclonal Antibody (CF594) [clone: HRB2/718]
Anti-ERBB2 Mouse Monoclonal Antibody (CF594) [clone: HRB2/718]
Supplier:  Biotium
Certificates
Anti-ERBB2 Mouse Monoclonal Antibody (CF594) [clone: HRB2/718]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75951-308
  • 75951-310
  • Antigen Symbol:
  • ERBB2
  • ERBB2
  • Antigen Name:
  • HER-2 / CD340
  • HER-2 / CD340
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • HRB2/718
  • HRB2/718
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 446352
  • Unigene ID: 446352
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications