Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). CK8 is primarily found in the non-squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas. It is absent in squamous cell carcinomas. Hepatocellular carcinomas are defined by the use of antibodies that recognize only cytokeratin 8 and 18. CK8 exists on several types of normal and neoplastic epithelia, including many ductal and glandular epithelia such as colon, stomach, small intestine, trachea, and esophagus as well as in transitional epithelium. Anti-CK8 does not react with skeletal muscle or nerve cells. Epithelioid sarcoma, chordoma, and adamantinoma show strong positivity corresponding to that of simple epithelia (with antibodies against CK8, CK18 and CK19). Reportedly, anti-CK8 is useful for the differentiation of lobular (ring-like, perinuclear) from ductal (peripheral-predominant) carcinoma of the breast.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.
Type: Primary
Antigen: KRT8
Clonality: Monoclonal
Clone: Clone C-43
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG's
Reactivity: Cow, Human, Pig, Rabbit, Sheep
- Catalog No:
- 75920-418
- 75920-416
- Antigen Symbol:
- KRT8
- KRT8
- Antigen Name:
- Cytokeratin 8
- Cytokeratin 8
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- C-43
- C-43
- Reactivity:
- Pig, Rabbit, Sheep, Human, Bovine
- Pig, Rabbit, Sheep, Human, Bovine
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 533782 , 708445
- Unigene ID: 533782 , 708445
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.1 mg/mL
- 0.1 mg/mL
- Storage Buffer:
- PBS/0.1% BSA/0.05% azide
- PBS/0.1% BSA/0.05% azide