Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CCNA2 Mouse Monoclonal Antibody (CF405S) [clone: E67]
Anti-CCNA2 Mouse Monoclonal Antibody (CF405S) [clone: E67]
Supplier:  Biotium
Certificates
Anti-CCNA2 Mouse Monoclonal Antibody (CF405S) [clone: E67]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75922-312
  • 75923-102
  • Antigen Symbol:
  • CCNA2
  • CCNA2
  • Antigen Name:
  • Cyclin A2
  • Cyclin A2
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • E67
  • E67
  • Reactivity:
  • Green monkey, Monkey, Human, Bovine, Mink, Mouse
  • Green monkey, Monkey, Human, Bovine, Mink, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 58974
  • Unigene ID: 58974
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together