Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ACTA2 Mouse Monoclonal Antibody (CF405S) [clone: Clone 1A4]
Anti-ACTA2 Mouse Monoclonal Antibody (CF405S) [clone: Clone 1A4]
Supplier:  Biotium
Certificates
Anti-ACTA2 Mouse Monoclonal Antibody (CF405S) [clone: Clone 1A4]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75981-368
  • 75981-370
  • Antigen Symbol:
  • ACTA2
  • ACTA2
  • Antigen Name:
  • Smooth Muscle Actin
  • Smooth Muscle Actin
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • 1A4
  • 1A4
  • Reactivity:
  • Rat, Mouse, Cat, Baboon, Pig, Rabbit, Monkey, Sheep, Goat, Chicken, Human, Bovine, Guinea pig, Dog
  • Rat, Mouse, Cat, Baboon, Pig, Rabbit, Monkey, Sheep, Goat, Chicken, Human, Bovine, Guinea pig, Dog
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 500483
  • Unigene ID: 500483
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together