Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-TNF Mouse Monoclonal Antibody (CF405S) [clone: TNF656]
 :  Biotium
 
Anti-TNF Mouse Monoclonal Antibody (CF405S) [clone: TNF656]
 :  Biotium

 

  • Catalog No:
  • 75977-028
  • 75977-030
  • Antigen Symbol:
  • TNF
  • TNF
  • Antigen Name:
  • TNF alpha
  • TNF alpha
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • TNF656
  • TNF656
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 241570
  • Unigene ID: 241570
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide