About this item
Fibroblast growth factor-1 (FGF-1), also designated acidic FGF, and fibroblast growth factor-2 (FGF-2), also designated basic FGF, are members of a family of growth factors that stimulate proliferation of cells of mesenchymal, epithelial and neuroectodermal origin. Additional members of the FGF family include the oncogenes FGF-3 (Int2) and FGF-4 (hst/Kaposi), FGF-5, FGF-6, FGF-7 (KGF), FGF-8 (AIGF), FGF-9 (GAF) and FGF-10 through FGF-23. Members of the FGF family share 30-55% amino acid sequence identity and similar gene structure, and are capable of transforming cultured cells when overexpressed in trans- fected cells. Cellular receptors for FGFs are members of a second multigene family, including four tyrosine kinases designated Flg (FGFR-1), Bek (FGFR-L), TKF and FGFR-3.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.
Type: Primary
Antigen: FGF23
Clonality: Monoclonal
Clone: Clone FGF23/638
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Specifications
- Catalog No:
- 75926-680
- 75926-482
- Antigen Symbol:
- FGF23
- FGF23
- Antigen Name:
- FGF23
- FGF23
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- FGF23/638
- FGF23/638
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 287370
- Unigene ID: 287370
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.1 mg/mL
- 0.1 mg/mL
- Storage Buffer:
- PBS/0.1% BSA/0.05% azide
- PBS/0.1% BSA/0.05% azide