Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-VWF Mouse Monoclonal Antibody [clone: 3E2D10]
Anti-VWF Mouse Monoclonal Antibody [clone: 3E2D10]
Catalog # 75979-572
Supplier:  Biotium
Anti-VWF Mouse Monoclonal Antibody [clone: 3E2D10]
Catalog # 75979-572
Supplier:  Biotium
Supplier Number:  BNUM0633-50

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    von Willebrand Factor
  • Antigen Symbol:
    VWF
  • Clonality:
    Monoclonal
  • Clone:
    3E2D10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 μL
  • Gene ID:
    Unigene ID: 440848
  • Storage Buffer:
    PBS, no BSA, no azide
  • Concentration:
    1 mg/mL
  • Cat. No.:
    75979-572
  • Supplier no.:
    BNUM0633-50

Specifications

About this item

von Willebrand Factor (vWF) is a multimeric glycoprotein that is found in endothelial cells, plasma and platelets. It acts as a carrier protein for Factor VIII and promotes platelet adhesion and aggregation. vWF undergoes a variety of posttranslational modifications that influence the affinity and availability for Factor VIII, including cleavage of the propeptide and formation of N-terminal disulfide bonds. This antibody helps to establish the endothelial nature of some lesions of disputed histogenesis, e.g. Kaposi's sarcoma and cardiac myxoma. It is widely used for differentiating vascular lesions from those of other tissue differentiation within a panel of other vascular markers although not all tumors of endothelial differentiation contain this antigen.

Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: Clone 3E2D10
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human