Anti-ESR1 Mouse Monoclonal Antibody [clone: ER506]
75924-892
: Biotium
- Antibody Type:Primary
- Antigen Name:Estrogen Receptor
- Antigen Symbol:ESR1
- Clonality:Monoclonal
- Clone:ER506
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:50 μL
- Gene ID:Unigene ID: 208124
- Storage Buffer:PBS, no BSA, no azide
- Concentration:1 mg/mL
- Cat. No.:75924-892
- Supplier no.:BNUM0506-50
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. It strongly stains nuclei of epithelial cells in breast carcinomas.
Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: Clone ER506
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human