Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-KRT77 & KRT76 Mouse Monoclonal Antibody (CF640R) [clone: AE-1 / AE-3]
Anti-KRT77 & KRT76 Mouse Monoclonal Antibody (CF640R) [clone: AE-1 / AE-3]
Supplier:  Biotium
Certificates
Anti-KRT77 & KRT76 Mouse Monoclonal Antibody (CF640R) [clone: AE-1 / AE-3]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75909-442
  • 75909-444
  • Antigen Symbol:
  • KRT77 & KRT76
  • KRT77 & KRT76
  • Antigen Name:
  • Cytokeratin
  • Cytokeratin
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • AE-1 / AE-3
  • AE-1 / AE-3
  • Reactivity:
  • Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
  • Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 334989 (KRT77) , 654392 (KRT76)
  • Unigene ID: 334989 (KRT77) , 654392 (KRT76)
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together