Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-CD7 Mouse Monoclonal Antibody [clone: B-F12]
 :  Biotium
 
Anti-CD7 Mouse Monoclonal Antibody [clone: B-F12]
 :  Biotium

 

  • Catalog No:
  • 75917-696
  • 75917-698
  • Antigen Symbol:
  • CD7
  • CD7
  • Antigen Name:
  • CD7
  • CD7
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • B-F12
  • B-F12
  • Reactivity:
  • Pig, Human, Guinea pig
  • Pig, Human, Guinea pig
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 186820
  • Unigene ID: 186820
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.2 mg/mL
  • 0.2 mg/mL
  • Storage Buffer:
  • PBS/0.05% BSA/0.05% azide
  • PBS/0.05% BSA/0.05% azide

 

Frequently Bought Together