Anti-INS Mouse Monoclonal Antibody [clone: E2-E3]
About this item
This antibody recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as beta-cell and insulinoma marker.
Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: Clone E2-E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Cow, Human, Mouse, Pig, Rabbit
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
This antibody recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as beta-cell and insulinoma marker.
Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: Clone E2-E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Cow, Human, Mouse, Pig, Rabbit
Specifications for Product Family
Specifications
- Catalog No:
- 75954-750
- 75954-752
- Antigen Symbol:
- INS
- INS
- Antigen Name:
- Insulin
- Insulin
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- E2-E3
- E2-E3
- Reactivity:
- Pig|Rabbit|Human|Bovine|Mouse
- Pig|Rabbit|Human|Bovine|Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 272259
- Unigene ID: 272259
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.2 mg/mL
- 0.2 mg/mL
- Storage Buffer:
- PBS/0.05% BSA/0.05% azide
- PBS/0.05% BSA/0.05% azide
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: BNUB0281-100Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.75954-750Antigen SymbolINSAntigen NameInsulinAntibody TypePrimaryClonalityMonoclonalConjugationUnconjugatedCloneE2-E3ReactivityPig|Rabbit|Human|Bovine|MouseHostMouseGene IDUnigene ID: 272259IsotypeIgG1 kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0.2 mg/mLStorage BufferPBS/0.05% BSA/0.05% azide - Supplier Number: BNUB0281-500Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.75954-752Antigen SymbolINSAntigen NameInsulinAntibody TypePrimaryClonalityMonoclonalConjugationUnconjugatedCloneE2-E3ReactivityPig|Rabbit|Human|Bovine|MouseHostMouseGene IDUnigene ID: 272259IsotypeIgG1 kappaImmunoChemistryYesImmunoFluorescenceYesConcentration0.2 mg/mLStorage BufferPBS/0.05% BSA/0.05% azide



