About this item
This MAb recognizes a 22 kDa protein, identified as Transgelin, also designated SM22-alpha. It may cross-react with SM22-beta. Transgelin is expressed abundantly in smooth muscle cells. The human transgelin gene encodes a 201 amino acid protein that contains nuclear factor-binding motifs known to regulate transcription in smooth muscle. During embryogenesis, transgelin is expressed in smooth, cardiac and skeletal muscle, but is restricted during late fetal development and adulthood to all vascular and visceral smooth muscle cells and low levels of expression in heart. Transgelin is down regulated in several transformed cell lines, indicating that a reduction of transgelin expression may be an early indicator of the onset of transformation. Transgelin also binds Actin, causing Actin fibers to gel within minutes of binding. Binding of transgelin to Actin occurs at a ratio of 1:6 Actin monomers.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Type: Primary
Antigen: TAGLN
Clonality: Monoclonal
Clone: Clone TAGLN/247
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Cow, Human, Mouse, Pig, Rabbit
Specifications
- Catalog No:
- 75977-652
- 75977-654
- Antigen Symbol:
- TAGLN
- TAGLN
- Antigen Name:
- Transgelin / SM22-alpha
- Transgelin / SM22-alpha
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Clone:
- TAGLN/247
- TAGLN/247
- Reactivity:
- Pig, Rabbit, Human, Bovine, Mouse
- Pig, Rabbit, Human, Bovine, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 410977
- Unigene ID: 410977
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.1 mg/mL
- 0.1 mg/mL
- Storage Buffer:
- PBS/0.1% BSA/0.05% azide
- PBS/0.1% BSA/0.05% azide