Anti-HLA-DPB1 Mouse Monoclonal Antibody [clone: BRA-FB6]
Catalog # 75961-044
Supplier: Biotium
Specifications
- Antibody Type:Primary
- Antigen Name:MHC II
- Antigen Symbol:HLA-DPB1
- Clonality:Monoclonal
- Clone:BRA-FB6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:50 μL
- Gene ID:Unigene ID: 347270
- Storage Buffer:PBS, no BSA, no azide
- Concentration:1 mg/mL
- Cat. No.:75961-044
- Supplier no.:BNUM0199-50
Specifications
About this item
This antibody recognizes a non-polymorphic determinant of DP-MHC class II. MHC class II antigens are transmembrane glycoproteins of non-covalently linked alpha (33-35 kDa) and beta (27-30 kDa) chains. It reportedly reacts with B- & non-T, non-B cell lines but not with T- and myeloid cell lines and leukemias. Differential expression of MHC class II antigens on fetal and adult lymphocytes, malignant B cells appears to reflect the stage of cell differentiation which may be useful in the study of lymphoproliferative disorders.
Type: Primary
Antigen: HLA-DPB1
Clonality: Monoclonal
Clone: Clone BRA-FB6
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human