Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-HLA-DPB1 Mouse Monoclonal Antibody (CF568) [clone: BRA-FB6]
Anti-HLA-DPB1 Mouse Monoclonal Antibody (CF568) [clone: BRA-FB6]
Supplier:  Biotium
Certificates
Anti-HLA-DPB1 Mouse Monoclonal Antibody (CF568) [clone: BRA-FB6]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75961-008
  • 75961-010
  • Antigen Symbol:
  • HLA-DPB1
  • HLA-DPB1
  • Antigen Name:
  • MHC II
  • MHC II
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • BRA-FB6
  • BRA-FB6
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 347270
  • Unigene ID: 347270
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications