Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-THY1 Mouse Monoclonal Antibody (CF488A) [clone: F15-42-1]
Anti-THY1 Mouse Monoclonal Antibody (CF488A) [clone: F15-42-1]
Supplier:  Biotium
Certificates
Anti-THY1 Mouse Monoclonal Antibody (CF488A) [clone: F15-42-1]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75908-660
  • 75908-658
  • Antigen Symbol:
  • THY1
  • THY1
  • Antigen Name:
  • CD90
  • CD90
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • F15-42-1
  • F15-42-1
  • Reactivity:
  • Monkey, Human
  • Monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 644697
  • Unigene ID: 644697
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications