About this item
This MAb reacts with a protein of 20-30 kDa, identified as PGP 9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UCHL-1). Initially, PGP 9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP 9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP 9.5 gene is believed to cause a form of Parkinson's disease.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.
Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: Clone 13C4
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Specifications
- Catalog No:
- 75969-752
- 75969-754
- Antigen Symbol:
- UCHL1
- UCHL1
- Antigen Name:
- pgp9.5
- pgp9.5
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- 13C4
- 13C4
- Reactivity:
- Zebrafish, Pig, Rabbit, Sheep, Human, Bovine, Guinea pig, Dog, Rat, Mouse
- Zebrafish, Pig, Rabbit, Sheep, Human, Bovine, Guinea pig, Dog, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 518731
- Unigene ID: 518731
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western Blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.1 mg/mL
- 0.1 mg/mL
- Storage Buffer:
- PBS/0.1% BSA/0.05% azide
- PBS/0.1% BSA/0.05% azide