Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-UCHL1 Mouse Monoclonal Antibody (CF405S) [clone: 13C4]
 :  Biotium
 
Anti-UCHL1 Mouse Monoclonal Antibody (CF405S) [clone: 13C4]
 :  Biotium

 

  • Catalog No:
  • 75969-730
  • 75969-728
  • Antigen Symbol:
  • UCHL1
  • UCHL1
  • Antigen Name:
  • pgp9.5
  • pgp9.5
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • 13C4
  • 13C4
  • Reactivity:
  • Zebrafish, Pig, Rabbit, Sheep, Human, Bovine, Guinea pig, Dog, Rat, Mouse
  • Zebrafish, Pig, Rabbit, Sheep, Human, Bovine, Guinea pig, Dog, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 518731
  • Unigene ID: 518731
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western Blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

 

Frequently Bought Together