Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Golgi Complex Mouse Monoclonal Antibody (CF488A) [clone: 371-4]
Anti-Golgi Complex Mouse Monoclonal Antibody (CF488A) [clone: 371-4]
Supplier:  Biotium
Certificates
Anti-Golgi Complex Mouse Monoclonal Antibody (CF488A) [clone: 371-4]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75948-974
  • 75948-976
  • Antigen Name:
  • Golgi Complex
  • Golgi Complex
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • 371-4
  • 371-4
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together