Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-H1 Mouse Monoclonal Antibody (CF594) [clone: AE-4]
 :  Biotium
 
Anti-H1 Mouse Monoclonal Antibody (CF594) [clone: AE-4]
 :  Biotium

 

  • Catalog No:
  • 75950-636
  • 75950-634
  • Antigen Symbol:
  • H1
  • H1
  • Antigen Name:
  • Histone H1
  • Histone H1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • AE-4
  • AE-4
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 226117 , 97358
  • Unigene ID: 226117 , 97358
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide