Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Granulocyte Marker Mouse Monoclonal Antibody (CF647) [clone: BM-2]
Anti-Granulocyte Marker Mouse Monoclonal Antibody (CF647) [clone: BM-2]
Supplier:  Biotium
Certificates
Anti-Granulocyte Marker Mouse Monoclonal Antibody (CF647) [clone: BM-2]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75949-296
  • 75949-294
  • Antigen Name:
  • Granulocyte Marker
  • Granulocyte Marker
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • BM-2
  • BM-2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together