About this item
This antibody recognizes a 183 kDa protein with DNA-binding characteristics, which is identified as a myeloid specific antigen. It reacts with myeloid precursor cells and granulocytes in bone marrow. Its antigen appears to be restricted to M2 and M3 acute myelogenous leukemia (AML) subtypes. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. This MAb reacts with early precursor and mature forms of human myeloid cells. It is useful in the identification of myelogenous leukemias, distinguishing granulocytic sarcomas from lymphoid malignancies and also in the study of differentiation and transformation of human myeloid cells. The biological function of this antigen is not clear, although it has been proposed that it may play a role in the differentiation of myeloid cells.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Type: Primary
Antigen: Granulocyte Marker
Clonality: Monoclonal
Clone: Clone BM-2
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- 75949-268
- 75949-266
- Antigen Name:
- Granulocyte Marker
- Granulocyte Marker
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Clone:
- BM-2
- BM-2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.1 mg/mL
- 0.1 mg/mL
- Storage Buffer:
- PBS/0.1% BSA/0.05% azide
- PBS/0.1% BSA/0.05% azide