Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF594) [clone: LN-3]
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF594) [clone: LN-3]
Supplier:  Biotium
Certificates
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF594) [clone: LN-3]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75951-476
  • 75951-478
  • Antigen Symbol:
  • HLA-DRB1
  • HLA-DRB1
  • Antigen Name:
  • HLA-DRB
  • HLA-DRB
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • LN-3
  • LN-3
  • Reactivity:
  • Monkey, Human
  • Monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 534322
  • Unigene ID: 534322
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications