About this item
This MAb reacts with a protein of 20-30 kDa, identified as PGP 9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UCHL-1). Initially, PGP 9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP 9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP 9.5 gene is believed to cause a form of Parkinson's disease.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®647 is a far-red fluorescent dye (Ex/Em 650/665 nm) with excellent brightness. It also is compatible with super-resolution imaging by STORM.
Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: Clone 31A3
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Cow, Human, Mouse, Pig, Rat
Specifications
- Catalog No:
- 75969-808
- 75969-810
- Antigen Symbol:
- UCHL1
- UCHL1
- Antigen Name:
- pgp9.5
- pgp9.5
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- 31A3
- 31A3
- Reactivity:
- Pig, Human, Bovine, Rat, Mouse
- Pig, Human, Bovine, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 518731
- Unigene ID: 518731
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.1 mg/mL
- 0.1 mg/mL
- Storage Buffer:
- PBS/0.1% BSA/0.05% azide
- PBS/0.1% BSA/0.05% azide