Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-TP53 Mouse Monoclonal Antibody (Biotin) [clone: BP53-12]
Anti-TP53 Mouse Monoclonal Antibody (Biotin) [clone: BP53-12]
Supplier:  Biotium
Certificates
Anti-TP53 Mouse Monoclonal Antibody (Biotin) [clone: BP53-12]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75969-082
  • 75969-080
  • Antigen Symbol:
  • TP53
  • TP53
  • Antigen Name:
  • Tumor protein p53
  • Tumor protein p53
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • BP53-12
  • BP53-12
  • Reactivity:
  • Hamster, Monkey, Chicken, Human, Dog
  • Hamster, Monkey, Chicken, Human, Dog
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 654481
  • Unigene ID: 654481
  • Isotype:
  • IgG2a
  • IgG2a
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications