Order Entry
Puerto Rico
ContactUsLinkComponent
TTC Solution 1%, HiMedia Laboratories
TTC Solution 1%, HiMedia Laboratories
Catalog # 89130-574
Supplier:  HiMedia
CAS Number:  
TTC Solution 1%, HiMedia Laboratories
Catalog # 89130-574
Supplier:  HiMedia
Supplier Number:  FD057-5VL
CAS Number:  

Specifications

  • Description:
    TTC Solution 1%
  • Cat. No.:
    89130-574
  • Volume:
    10 ml per vial
  • Supplier no.:
    FD057-5VL

Specifications

About this item

Filter sterilized solution of 2,3,5-triphenyl tetrazolium chloride (TTC) used for the detection of microbial growth by means of TTC reduction.