Ethanol Spray Cleaner, ITW Texwipe®
: Texwipe
Sterile, ready-to-use 70% denatured ethanol (EtOH) solution designed especially for cleanroom use. Filtered to 0.2µm, individually double-bagged and gamma-irradiated to ensure sterility.
- Surface cleaning and residue removal
- Use on gloved hands in sterile suites
- Wipe down for pass-through to controlled environments
- Ideally suited for use with sterile cleanroom wipers
- 70% Denatured ethanol
: Packaged in a ISO Class 5 cleanroom.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.