Specifications
- Description:Bioprofile Control Level 5
- Cat. No.:101415-110
- Supplier no.:24619
Specifications
About this item
An assayed aqueous quality control material intended for monitoring the measurement of glucose, lactate, glutamine and glutamate.
Ingredients: These controls are formulated from a buffered aqueous solution, each with a known pH and containing known levels of glutamine and glutamate. The volume of each ampule is 1.7 mL.