Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Cell Staining Buffer, Biolegend

Cell Staining Buffer, Biolegend

Cell Staining Buffer, 500 mL
Catalog # 420201-BL
Supplier:   Biolegend
Supplier #:  420201
Cell Staining Buffer, Biolegend
Catalog # 420201-BL
Supplier:  Biolegend
CAS Number:  
Cell Staining Buffer, Biolegend

Cell Staining Buffer, 500 mL

CAS Number:  
Price
$124.00
Log in for availability
Each (500ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cell staining buffer is an antibody diluent and cell wash buffer optimized for use in immunofluorescent staining of viable or fixed single cell suspensions.

Cell Staining Buffer contains bovine calf serum as a protein carrier to reduce non-specific binding of antibodies and fluorochrome reagents to target cells. It also contains a metabolic inhibitor, sodium azide (NaN3), to inhibit patching and capping of cell surface antigens. To prevent interference in biotin/avidin indirect staining protocols, cell staining buffer is formulated without biotin.

Specifications

Specifications

  • Description:
    Cell Staining Buffer
  • Size:
    500 mL
  • Application:
    Staining of single Cell Suspensions
  • Cat. No.:
    420201-BL
  • Storage Temperature:
    2 °C – 8 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.