Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
pH Probe with Flat Surface, Sper Scientific

pH Probe with Flat Surface, Sper Scientific

pH probe with flat surface, 1 m (3.3 ft.), BNC Connector
Catalog # 76176-940
Supplier:   Sper Scientific
Supplier #:  840051
pH Probe with Flat Surface, Sper Scientific
Catalog # 76176-940
Supplier:  Sper Scientific
pH Probe with Flat Surface, Sper Scientific
pH Probe with Flat Surface, Sper Scientific

pH probe with flat surface, 1 m (3.3 ft.), BNC Connector

Price
$271.43
Log in for availability
Each
Product Details & Documents

pH probe is useful for reading the pH of small amounts of liquid, food products, and other surface pH measurements.

  • Easy to clean
  • Features a single-junction electrode for general-purpose applications

It features flat surface and is commonly used in testing surface measurements such as cheese, meats, small liquid samples, paper, soil, or skin.

Delivery information: Probe comes with a 40" cable and a standard BNC connector.

Specifications
  • Description:
    pH probe with flat surface
  • Cable length:
    1 m (3.3 ft.)
  • Connector:
    BNC Connector
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.