Specifications
- Description:ADP-Glo Kinase Assay + TBK1 Kinase Enzyme System, 1 each
- Size:1 each
- Storage Temperature:°COMPONENTS AT DIFFERENT TEMPS
- Cat. No.:PAV8291
- Supplier no.:V8291
Specifications
About this item
Recombinant full-length human TBK1 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. TBK1, also known as NAK or NFkB-activating kinase, is an upstream protein kinase that can phosphorylate and activate the IkB kinases.
- Easily Screen and Profile TBK1 Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human TBK1 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. TBK1, also known as NAK or NFkB-activating kinase, is an upstream protein kinase that can phosphorylate and activate the IkB kinases. Activation of IkB kinases allows the phosphorylation of IkB protein, which is then degraded via the ubiquitination pathway. This mechanism allows the activation of the NFkB transcriptional complex. TBK1 is a specific upstream regulator of IkB kinases and also can interact with the IkB protein TANK. TBK1 is a component of the virus-activated kinase that phosphorylate IRF3 and IRF7, allowing their dimerization and translocation to the nucleus, where they induce transcription of interferon. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: TBK1, 10ug (Human, recombinant full-length). MW: ~105kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. TBK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/29110/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.