Order Entry
Puerto Rico
ContactUsLinkComponent
ADP-Glo Kinase Assay + PKD2 Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + PKD2 Kinase Enzyme System, 1 each, Promega
Catalog # PAV4043
CAS Number:  
undefined
ADP-Glo Kinase Assay + PKD2 Kinase Enzyme System, 1 each, Promega
Catalog # PAV4043
Supplier Number:  V4043
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + PKD2 Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV4043
  • Supplier no.:
    V4043

Specifications

About this item

Recombinant full-length human PKD2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKD2 is a novel phorbol ester- and growth factor-stimulated serine/threonine kinase.

  • Easily Screen and Profile PKD2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant full-length human PKD2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKD2 is a novel phorbol ester- and growth factor-stimulated serine/threonine kinase that contains two cysteine-rich motifs at the N terminus, a pleckstrin homology domain and a catalytic domain (1). It exhibits the strongest homology to the serine/threonine protein kinases PKD/PKC mu and PKC nu. The PKD family of enzymes has been implicated in very diverse cellular functions, including Golgi organization and plasma membrane directed transport, metastasis, immune responses, apoptosis and cell proliferation (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PKD2, 10ug (Human, recombinant full-length). MW: ~130kDa. Substrate: CREBtide (KRREILSRRPSYR); derived from human CREB1 isoform A (amino acids 109-121). Other: Reaction Buffer, DTT. PKD2 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/25865/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.