Specifications
- Description:ADP-Glo Kinase Assay + PDGFR alpha (T674I) Kinase Enzyme System, 1 each
- Size:1 each
- Storage Temperature:°COMPONENTS AT DIFFERENT TEMPS
- Cat. No.:PAV4487
- Supplier no.:V4487
Specifications
About this item
Recombinant human PDGFRα (Y674I) (550-end) point mutant was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
- Easily Screen and Profile PDGFRα (T674I) Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human PDGFRα (Y674I) (550-end) point mutant was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PDGFRα (platelet-derived growth factor receptor α) is a member of the PDGFR family of membrane receptors with intrinsic tyrosine kinase activity. Aberrant expression of PDGFRα has been linked to developmental abnormalities in vertebrate models and implicated in multiple disease states in humans. There is widespread expression of PDGFRα in renal cell types involved in fibrotic and sclerosing processes. PDGF and its receptor PDGFRα are inducers of fibrosis in the repair phase of inflammatory bowel disease, and they may also be involved in the active inflammatory phase. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PDGFRα (D842V), 10microg (Human, recombinant; amino acids 550-end). MW: ~95kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, MnCl2, DTT. PDGFRα (T674I) NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5156/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.