Order Entry
Puerto Rico
ContactUsLinkComponent
ADP-Glo Kinase Assay + MELK Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + MELK Kinase Enzyme System, 1 each, Promega
Catalog # PAV4151
CAS Number:  
undefined
ADP-Glo Kinase Assay + MELK Kinase Enzyme System, 1 each, Promega
Catalog # PAV4151
Supplier Number:  V4151
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ADP-Glo Kinase Assay + MELK Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV4151
  • Supplier no.:
    V4151

Specifications

About this item

Recombinant human MELK (1-340) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MELK or maternal embryonic leucine zipper kinase is a member of the Snf1/AMPK kinase family.

  • Easily Screen and Profile MELK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant human MELK (1-340) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MELK or maternal embryonic leucine zipper kinase is a member of the Snf1/AMPK kinase family. MELK is a key regulator of the proliferation of malignant brain tumors including their stem cells (1). MELK transcript abundance correlates with malignancy grade in human astrocytomas and represents a therapeutic target for the management of the most frequent brain tumors in adult and children. MELK also plays a role in mammary carcinogenesis through inhibition of the pro-apoptotic function of Bcl-GL (2). Therefore, the kinase activity of MELK could be a promising molecular target for development of therapy for patients with breast cancers. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: MELK, 10ug (Human, recombinant; amino acids 1-340). MW: ~61kDa. Substrate: ZIPtide (KKLNRTLSFAEPG). Other: Reaction Buffer, DTT. MELK NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/9833/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.