Order Entry
Puerto Rico
ContactUsLinkComponent
ADP-Glo Kinase Assay + HPK1 Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + HPK1 Kinase Enzyme System, 1 each, Promega
Catalog # PAV4099
CAS Number:  
undefined
ADP-Glo Kinase Assay + HPK1 Kinase Enzyme System, 1 each, Promega
Catalog # PAV4099
Supplier Number:  V4099
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + HPK1 Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV4099
  • Supplier no.:
    V4099

Specifications

About this item

Recombinant human HPK1 (1-346) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. HPK1 or Hematopoietic progenitor kinase 1 is a hematopoietic cell-restricted member of the Ste20 serine/threonine kinase super family.

  • Easily Screen and Profile HPK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant human HPK1 (1-346) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. HPK1 or Hematopoietic progenitor kinase 1 is a hematopoietic cell-restricted member of the Ste20 serine/threonine kinase super family. HPK1 is also known as mitogen-activated protein kinase kinase kinase kinase 1 (MAP4K1). HPK1 is a tissue-specific upstream activator of the MEKK/JNK/SAPK signaling pathway (1). HPK1 diminishes T cell receptor (TCR) signaling activity and T cell proliferation by phosphorylating the adaptor protein SLP-76 (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: HPK1, 10ug (Human, recombinant; amino acids 1-346). MW: ~65kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. HPK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/11184/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.