Order Entry
Puerto Rico
ContactUsLinkComponent
 
ADP-Glo Kinase Assay + FYN A Kinase Enzyme System, 1 each, Promega
Catalog # PAV9341
CAS Number:  
undefined
ADP-Glo Kinase Assay + FYN A Kinase Enzyme System, 1 each, Promega
Catalog # PAV9341
Supplier Number:  V9341
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + FYN A Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV9341
  • Supplier no.:
    V9341

Specifications

About this item

Full-length recombinant human FYN A was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FYN A (Fyn isoform A) is a member of the SRC tyrosine kinase oncogene family showing high homology to YES1, FGR and SRC.

  • Easily Screen and Profile FYN A Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Full-length recombinant human FYN A was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FYN A (Fyn isoform A) is a member of the SRC tyrosine kinase oncogene family showing high homology to YES1, FGR and SRC. FYN has been shown to phosphorylate DAB1, an intracellular adaptor protein that interacts with amyloid precursor protein (APP) and apoE receptor 2 (apoEr2). The interaction of FYN and DAB1 regulates the phosphorylation, trafficking and processing of APP and apoEr2. FYN expression has been shown to be significantly increased in Chronic Myelogenous Leukemia (CML). Knockdown of FYN with shRNA slows leukemia cell growth, inhibits clonogenicity and leads to increased sensitivity to imatinib. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: FYN A, 10ug (Human, recombinant full-length). MW: ~85kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT. FYN A NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/2534/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.