Order Entry
Puerto Rico
ContactUsLinkComponent
ADP-Glo Kinase Assay + FMS Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + FMS Kinase Enzyme System, 1 each, Promega
Catalog # PAV4023
CAS Number:  
undefined
ADP-Glo Kinase Assay + FMS Kinase Enzyme System, 1 each, Promega
Catalog # PAV4023
Supplier Number:  V4023
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + FMS Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV4023
  • Supplier no.:
    V4023

Specifications

About this item

Recombinant human FMS (539-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FMS is a proto-oncogene that encodes the tyrosine kinase transmembrane receptor for colony stimulating factor 1 receptor (CSF1R).

  • Easily Screen and Profile FMS Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant human FMS (539-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FMS is a proto-oncogene that encodes the tyrosine kinase transmembrane receptor for colony stimulating factor 1 receptor (CSF1R). FMS is homodimeric, contains a so-called kinase insert domain and is a member of the CSF1R/PDGF receptor family of tyrosine-protein kinases. FMS mediates most if not all of the biological effects of CSF1, which control the production, differentiation and cell function the monocyte/macrophage lineage (1). Mutations in FMS have been associated with providing sustained signals for cell growth and a predisposition to myeloid malignancy (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: FMS, 10ug (Human, recombinant; amino acids 539-end). MW: ~76kDa. Substrate: Poly (4:1 Glu, Tyr) peptide. Other: Reaction Buffer, DTT, MnCl2. FMS NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/1436/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.