Order Entry
Puerto Rico
ContactUsLinkComponent
Wizard Plus Megapreps DNA Purification System, 5 preps, Promega®
Wizard Plus Megapreps DNA Purification System, 5 preps, Promega®
  PAA7300
 :  
undefined
Wizard Plus Megapreps DNA Purification System, 5 preps, Promega®
  PAA7300
 :  A7300
 :  

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mega
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Wizard Plus Megapreps DNA Purification System, 5 preps
  • Size:
    5 preps
  • Sample Size:
    600 mL–1 L of culture
  • Description:
    Wizard Plus Megapreps DNA Purification System
  • Yield:
    >1 mg
  • Cat. No.:
    PAA7300
  • Supplier no.:
    A7300

 

 

A simple, reliable, silica-resin-based method for rapid isolation of plasmid DNA from 1 liter cultures.

  • More than 1mg Plasmid from 1L Cultures
  • 3-hour protocol
  • Resin-based batch column method
  • No organic extractions

Wizard Plus Megapreps DNA Purification System provides a simple and rapid method for large-scale purifications of plasmid DNA that eliminates the need for cesium chloride:ethidium bromide gradient centrifugation. Use of this system requires only a centrifuge, a vacuum source and a vacuum manifold. The system yields greater than one milligram of high-copy-number plasmid DNA (200-20,000bp) from a 1,000ml culture in less than three hours. The purified DNA is eluted in Nuclease-Free Water (Cat.# P1193) or TE buffer and can be used directly for DNA sequencing and restriction digestion without further manipulation. The DNA also can be used for in vitro transcription reactions supplemented with a ribonuclease inhibitor such as Recombinant RNasin Ribonuclease Inhibitor (Cat.# N2511).