Order Entry
Puerto Rico
ContactUsLinkComponent
Wizard DNA Clean-Up System, 100 preps, Promega
Wizard DNA Clean-Up System, 100 preps, Promega
Catalog # PAA7280
CAS Number:  
undefined
Wizard DNA Clean-Up System, 100 preps, Promega
Catalog # PAA7280
Supplier Number:  A7280
CAS Number:  

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    Wizard DNA Clean-Up System, 100 preps
  • Size:
    100 preps
  • Sample Size:
    50 ng–16 µg
  • Description:
    Wizard DNA Clean-Up System
  • Yield:
    80–90%
  • Cat. No.:
    PAA7280
  • Supplier no.:
    A7280

Specifications

About this item

A simple and effective way to purify linear and circular DNA (200-50,000bp) from many molecular biology reactions.

  • Purifies DNA (200-50,000bp) from Molecular Biology Reactions
  • Results in 15 minutes or less
  • No phenol extractions or ethanol precipitations

The Wizard DNA Clean-Up System provides a simple and effective way to purify linear and circular DNA (200-50,000bp) from many molecular biology reactions. Using a quick batch-column procedure, the entire process can be completed in 15 minutes or less with no organic extractions or ethanol precipitations. DNA is eluted in water or TE buffer, ready for use. Multiple preps may be processed easily at one time with the Vac-Man Laboratory Vacuum Manifold (Cat.# A7231).