Order Entry
Puerto Rico
ContactUsLinkComponent
Wizard Plus Maxipreps DNA Purification System, 10 preps, Promega®
Wizard Plus Maxipreps DNA Purification System, 10 preps, Promega®
Catalog # PAA7270
CAS Number:  
undefined
Wizard Plus Maxipreps DNA Purification System, 10 preps, Promega®
Catalog # PAA7270
Supplier Number:  A7270
CAS Number:  

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Maxi
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Wizard Plus Maxipreps DNA Purification System, 10 preps
  • Size:
    10 preps
  • Sample Size:
    100–500 mL of culture
  • Description:
    Wizard Plus Maxipreps DNA Purification System
  • Yield:
    300 µg–1 mg
  • Cat. No.:
    PAA7270
  • Supplier no.:
    A7270

Specifications

About this item

A simple, reliable, silica-resin-based method for isolation of plasmid DNA on a maxiprep scale. No organic extraction or ethanol precipitation required. Yields 300ug-1mg from 100-500ml culture.

  • 300µg-1mg Plasmid DNA from 100-500ml Cultures
  • 3-hour protocol
  • Resin-based batch-binding and column-washing method
  • No organic extractions

The Wizard Plus Maxipreps DNA Purification System provides a simple and rapid resin-based batch column method for purification of plasmid DNA that eliminates the need for cesium chloride:ethidium bromide gradient centrifugation. Use of this system requires only a centrifuge, a vacuum source and a vacuum manifold. This system typically yields 300ug-1mg of high-copy-number plasmid DNA (200-20,000bp) from a 100-500ml culture in less than three hours. The purified DNA is eluted in Nuclease-Free Water (Cat.# P1193) and can be used directly for DNA sequencing and restriction digestion without further manipulation. The DNA also can be used for in vitro transcription reactions supplemented with a ribonuclease inhibitor such as Recombinant RNasin Ribonuclease Inhibitor (Cat.# N2511).