Order Entry
Puerto Rico
ContactUsLinkComponent
ADP-Glo Kinase Assay + FGFR1 Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + FGFR1 Kinase Enzyme System, 1 each, Promega
Catalog # PAV9321
CAS Number:  
undefined
ADP-Glo Kinase Assay + FGFR1 Kinase Enzyme System, 1 each, Promega
Catalog # PAV9321
Supplier Number:  V9321
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + FGFR1 Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV9321
  • Supplier no.:
    V9321

Specifications

About this item

Recombinant human FGFR1 (amino acids 399-822) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FGFR1 (also known as FLT2) is a member of the Fibroblast Growth Factor Receptor family.

  • Easily Screen and Profile FGFR1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant human FGFR1 (amino acids 399-822) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FGFR1 (also known as FLT2) is a member of the Fibroblast Growth Factor Receptor family that constitutes a family of four membrane-spanning tyrosine kinases (FGFR1-4), which serve as high-affinity receptors for 17 growth factors (FGF1-17). The FGF Receptor family plays an important role in multiple biological processes, including mesoderm induction and patterning, cell growth and migration, organ formation and bone growth. FGFR1 is alternatively spliced, generating multiple splice variants that are differentially expressed during embryo development and in the adult body. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: FGFR1, 10ug (Human, recombinant; amino acids 399-822). MW: ~73kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT. FGFR1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/2260/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.