Order Entry
Puerto Rico
ContactUsLinkComponent
ADP-Glo Kinase Assay + ERK2 Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + ERK2 Kinase Enzyme System, 1 each, Promega
Catalog # PAV9291
CAS Number:  
undefined
ADP-Glo Kinase Assay + ERK2 Kinase Enzyme System, 1 each, Promega
Catalog # PAV9291
Supplier Number:  V9291
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + ERK2 Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV9291
  • Supplier no.:
    V9291

Specifications

About this item

Recombinant full-length human ERK2 was expressed by E. coli cells using an N-terminal GST tag and activated by MEK1 in vitro. ERK2 is a protein serine/threonine kinase that is a member of the extracellular signal-regulated kinases (ERKs).

  • Easily Screen and Profile ERK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant full-length human ERK2 was expressed by E. coli cells using an N-terminal GST tag and activated by MEK1 in vitro. ERK2 is a protein serine/threonine kinase that is a member of the extracellular signal-regulated kinases (ERKs), which are activated in response to numerous growth factors and cytokines. Activation of ERK2 requires both tyrosine and threonine phosphorylation that is mediated by MEK. ERK2 is ubiquitously distributed in tissues with the highest expression in heart, brain and spinal cord. Activated ERK2 translocates into the nucleus where it phosphorylates various transcription factors (e.g., Elk-1, c-Myc, c-Jun, c-Fos and C/EBP beta). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: ERK2, 10ug (Human, recombinant full-length). MW: ~68kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. ERK2 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5594/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.