Order Entry
Puerto Rico
ContactUsLinkComponent
ADP-Glo Kinase Assay + EIF2AK2 Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + EIF2AK2 Kinase Enzyme System, 1 each, Promega
Catalog # PAV5329
CAS Number:  
undefined
ADP-Glo Kinase Assay + EIF2AK2 Kinase Enzyme System, 1 each, Promega
Catalog # PAV5329
Supplier Number:  V5329
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + EIF2AK2 Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV5329
  • Supplier no.:
    V5329

Specifications

About this item

Recombinant human EIF2AK2 (252-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EIF2AK2 (also known as double-stranded RNA-activated protein kinase) is shown to be involved in HIV/gp120-associated neurodegeneration.

  • Easily Screen and Profile EIF2AK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant human EIF2AK2 (252-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EIF2AK2 (also known as double-stranded RNA-activated protein kinase) is a protein kinase shown to be involved in HIV/gp120-associated neurodegeneration. EIF2AK2 acts as a critical mediator of gp120 neurotoxicity and is a substrate for a family of protein kinases that respond to various forms of environmental stress. Activation of EIF2AK2 leads to its autophosphorylation and then phosphorylation of its natural substrate, the alpha subunit of eukaryotic protein synthesis initiation factor-2. EIF2AK2 plays a critical role in mRNA translation, cell proliferation and apoptosis. A novel cross-talk event between the EIF2AKs and p53 has been shown that has implications in cell proliferation and tumorigenesis. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: EIF2AK2, 10microg (Human, recombinant; amino acids 252-end). MW: ~64kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. EIF2AK2 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5610/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.