Order Entry
Puerto Rico
ContactUsLinkComponent
Wizard PCR Preps DNA Purification System, 50 preps, Promega
Wizard PCR Preps DNA Purification System, 50 preps, Promega
Catalog # PAA7170
CAS Number:  
undefined
Wizard PCR Preps DNA Purification System, 50 preps, Promega
Catalog # PAA7170
Supplier Number:  A7170
CAS Number:  

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    Wizard PCR Preps DNA Purification System, 50 preps
  • Size:
    50 preps
  • Sample Size:
    50 ng–16 µg of DNA
  • Description:
    Wizard PCR Preps DNA Purification System
  • Yield:
    80–90% of PCR or 60–80% of DNA from gel
  • Cat. No.:
    PAA7170
  • Supplier no.:
    A7170

Specifications

About this item

A 15-minute column purification method for purifying PCR-amplified DNA away from contaminants, including primer-dimers and amplification primers. Can also be used to purify DNA fragments from agarose gels.

  • Purifies PCR Products Directly from Reactions
  • 15-minute protocol
  • Purify PCR DNA from reactions or gel slices

The Wizard PCR Preps DNA Purification System provides a simple, reliable way to purify double-stranded PCR-amplified DNA. Using the 15-minute batch column purification method, PCR products are effectively separated from contaminants, including primer-dimers and amplification primers. This system also can be used to purify DNA fragments from agarose gels. The DNA can be eluted in water or TE buffer, free of salts or macromolecular contaminants. Multiple PCR Preps may be processed easily at one time with the Vac-Man Laboratory Vacuum Manifold (Cat.# A7231).